Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Lady Beetles
SCI_ED

Well-known predator of aphids.

Specifications

Product Type Lady Beetle
For Use With (Application) Well known predator of aphids, often used for biological pest control.
Scientific Name Hippodamia convergens
Storage Requirements 18°C
View More Specs

Alert:

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000


Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
S07642ND
SCI_ED
View Documents
Carolina Biological SupplySupplier Diversity Partner
144222
100/Container N/A N/A
This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-955-1177. Provide the NEED BY DATE to the customer service agent.
Description

Description

  • Hippodamia convergens
  • Often used for biological pest control

Specifications

Specifications

Lady Beetle
Well known predator of aphids, often used for biological pest control.
Hippodamia convergens
18°C
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit