Learn More
Carolina™ Mealworms
SCI_ED
The adult mealworm lays hundreds of eggs and is the best stage to start a balanced culture. Larvae are ideal food for lizards, salamanders, turtles, and frogs.
Specifications
Scientific Name Tenebrio
Storage Requirements 25° to 30°C
View More Specs
Alert:
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Products 2
Catalog Number Mfr. No. Product Type Quantity For Use With (Application) Includes Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Quantity For Use With (Application) Includes Price Quantity & Availability  
S07275CND
SCI_ED
View Documents
Carolina Biological SupplySupplier Diversity Partner
TF144274LC
Mealworm Coupon 100/Container Ideal food for lizards, salamanders, turtles and frogs. Care instructions N/A N/A
This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-955-1177. Provide the NEED BY DATE to the customer service agent.
S07275ND
SCI_ED
View Documents
Carolina Biological SupplySupplier Diversity Partner
144274
Mealworm 100/Container Ideal food for lizards, salamanders, turtles and frogs. Care instructions N/A N/A
This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-955-1177. Provide the NEED BY DATE to the customer service agent.
Description
Description
  • Tenebrio
  • Pupae metamorphose into adults in about two to four weeks
  • With instructions
Note: Prepaid coupons allow you to order your organisms in advance. We'll send a coupon(s) that you later redeem by phone, fax, Online web form, or mail. To ensure prompt arrival of material, coupons must be redeemed at least 2 weeks prior to requested delivery date.
Specifications
Specifications
Tenebrio
25° to 30°C