Carolina Men's Style 6549 Athletic Steel Toe Hiker

Manufacturer: Carolina Shoe Company 6549M115

 View more versions of this product

Catalog No. 19-150-0813

  Check Availability
Add to Cart

    Specifications

    Footbed Material EVA
    Certifications/Compliance ASTM, M I/75, C/75
    Eyelets Material Non Metallic
    For Use With (Application) Athletically inspired shoe that's built for work
    Gender Male
    Liner Material Nylon Mesh Fabric
    Shank Material Steel
    Sole Features Cement (flexible)
    Style Athletic
    Toe Material Steel
    Tread Style Aggressive
    Waterproof No
    Color Brown
    Material Nubuck Leather
    Size 11.5M

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit