Learn More
Carolina Men's Style 6549 Athletic Steel Toe Hiker
Manufacturer:  Carolina Shoe Company 6549M090
 View more versions of this product
Catalog No. 19-150-0808
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
ASTM, M I/75, C/75
EVA
Male
Steel
Athletic
Aggressive
Brown
9M
Non Metallic
Athletically inspired shoe that's built for work
Nylon Mesh Fabric
AeroTrek™ Rubber outsole, slip resistant
Steel
No
Nubuck Leather