Learn More
Carolina™ Plaque Demonstration Kit A with Prepaid Coupon Products for Science Education
Designed to initiate and teach handling of plaque-forming bacteriophages.
Manufacturer:  Carolina™ 124310
Catalog No. S21832CND
This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-955-1177. Provide the NEED BY DATE to the customer service agent.
Alert:
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Includes: Prepaid coupon
Description
Description
  • Allows serial dilution and production of plaques by agar-layer technique
  • Suitable for individual projects or class exercises
  • Instructions are written for participation by four students
  • Uses Coliphages T4 and Mutant T4r
Note: Refrigerate coliphage until use; do not freeze. Keep bacteria culture and agar at room temperature
Specifications
Specifications
Serial dilution and the production of plaques by the agar-layer technique
Refrigerate coliphage until use; do not freeze. Keep bacteria culture and agar at room temperature.
Prepaid coupon
Education
1 Kit
Plaque Demonstration Kit A