Carolina PVC Gloves

Manufacturer: Carolina Glove CompanySupplier Diversity Partner Y010

 View more versions of this product

Catalog No. 19-052952

  Check Availability
Add to Cart

    Specifications

    Cleaning Procedure Wash in warm Water, air dry and no Bleach
    Exterior Finish Smooth
    Glove Cuff Style Gauntlet
    For Use With (Application) chemical, construction, fishing, mining, Oil, Gas drilling, painting and refining industries
    Length (English) 10 in.
    Length (Metric) 25.4cm
    Material PVC
    Liner Material Cotton Jersey
    Color Yellow

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit