Learn More
Carolina™ Ramhorn Snails Products for Science Education
The ramhorn snail is the most popular aquarium snail.
Manufacturer:  Carolina Biological SupplySupplier Diversity Partner 141230
 View more versions of this product
Catalog No. S07607ND
This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-955-1177. Provide the NEED BY DATE to the customer service agent.
Alert:
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Description
Description
Their shells range from translucent to various shades of brown to nearly black.
  • Available year-round
  • Excellent for keeping aquariums clean
  • With instructions
Specifications
Specifications
Excellent for keeping aquariums clean
12 Pack
Ramhorn Snail
18° to 25°C
Instructions