Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Shoe Men's 6 in. Waterproof Carbon Composite Toe Hiker Boots

Manufacturer:  Carolina™ CA4525D140

 View more versions of this product

Catalog No. 19-152-212


Add to cart

Specifications

Specifications

Safety
Non-metallic
6 in.
2.54 cm
Nylon
EVA midsole, 4 x 4 Rubber outsole, Pillow Cushion™ insole, slip-resistant
Carbon Composite Fiber
Yes
Brown
AG7™ Polyurethane
1 in.
Leather
14D
Hiker
Aggressive
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit