Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina Shoe Men's 6 in. Waterproof Carbon Composite Toe Hiker Boots

Manufacturer:  Carolina Shoe Company CA4525D100

 View more versions of this product

Catalog No. 19-152-206


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Non Metallic
Male
6 in.
Nylon
Hiker
Aggressive
Dark Brown
10D
AG7™ Polyurethane
1 in.
2.54cm
EVA midsole, 4 x 4 Rubber outsole, Pillow Cushion™ insole, slip resistant
Carbon composite Fiber
Yes
Leather
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit