Carolina Shoe Waterproof Composite Ranch Wellington Work Boots

Manufacturer: Carolina Shoe Company CA2520D115

 View more versions of this product

Catalog No. 19-089-129

  Check Availability
Add to Cart

    Specifications

    Gender Male
    Sole Features Cement (flexible)
    Waterproof Yes
    Color Brown
    Size 11.5D
    Certifications/Compliance ASTM
    Footbed Material AG7™ Polyurethane
    For Use With (Application) General purpose work
    Height (English) 10 in.
    Shank Material Nylon
    Style Pull-on Wellington
    Toe Style Composite
    Tread Style Smooth minimal
    Material Folklore Leather

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit