Carolina Shoe Women's ESD Safety-Toe Athletic Shoes
With mesh comfort lining
Manufacturer: Carolina Shoe Company CA9513W110
 View more versions of this product
Catalog No. 19-152-269
  Check Availability
Add to Cart
    Specifications
    Footbed Material EVA
    Gender Female
    Liner Material Mesh
    Shank Material Non Metallic
    Sole Features Cement (flexible)
    Toe Style Toe cap
    Color Tan/Black
    Size 11W
    Certifications/Compliance ASTM F2413-05
    Material Leather
  • Description & Specifications
Related Products
Product Content Feedback