Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Slime Mold Cultures
SCI_ED

Slime mold is a broad term describing some organisms that use spores to reproduce.

$16.25

Specifications

Quantity Each
Includes Carolina™ techniques for studying bacteria and fungi manual
Sufficient For Class of 30 students
Temperature (Metric) Range 25°C
View More Specs

Alert:

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000


Includes:

  • Enough materials for 30 students


Products 1
Catalog Number Mfr. No. Product Type Media Type Packaging Scientific Name Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Media Type Packaging Scientific Name Price Quantity & Availability  
S07213ND
SCI_ED
View Documents
Carolina Biological SupplySupplier Diversity Partner
156193
Physarum polycephalum plasmodium Culture 2% Agar with old fashioned Quaker™ oats Plate Physarum polycephalum plasmodium
Each for $16.25
Add to cart
 
Description

Description

  • Myxomycetes
  • Note: To ensure you order enough living materials for the number of students in your class, order one culture for up to 30 students, two cultures for 31 to 60 students, three cultures for 61 to 90 students, etc.

‡When you receive your Phyusarum culture, remove the sealing tape from the Petri dish and sprinkle a few grains of oat flakes (rolled oats) along the edges of the culture growth. Keep the Petri dish at room temperature (68 to 72°F, 20 to 22°C) in indirect light until ready to use. When Physarum reaches the full perimeter of the Petri dish, set up a new culture to keep your slime mold at its best.

Specifications

Specifications

Each
Carolina™ techniques for studying bacteria and fungi manual
Class of 30 students
25°C
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit