Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Tree Frogs
SCI_ED

Excellent for the terrarium.

Specifications

Product Type Tree Frog
For Use With (Application) Excellent for the terrarium
Includes Instructions
Scientific Name Hyla
Storage Requirements 23° to 29°C
View More Specs

Alert:

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000


Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
S07739ND
SCI_ED
View Documents
Carolina Biological SupplySupplier Diversity Partner
146720
3/Container N/A N/A
This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-955-1177. Provide the NEED BY DATE to the customer service agent.
Description

Description

  • Hyla
  • Species not guaranteed
  • With complete care instructions

Specifications

Specifications

Tree Frog
Instructions
23° to 29°C
Excellent for the terrarium
Hyla
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit