Learn More
Carolina Tropical Fish Set
Tropical fish such as bettas, gouramis, chinese algae eaters, guppies, and algae-eating shrimp are excellent choices for the classroom aquarium.
Manufacturer: Carolina Biological Supply 145470
Alert:
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Includes:
- 12 tropical fish (at least three different types)
- Instructions
Description
All tropical species are hand-selected for size and health, for years of enjoyment.
- Aquaria for these fish require a heater to maintain proper water temperature (75 to 80°F)
- Suitable for a community tank
- For freshwater aquaria
Specifications
Excellent choices for the classroom aquarium, all tropical species are hand selected for size and health. | |
Each | |
Tropical Fish Set |
23.88° to 26.66°C | |
12 tropical fish (at least three different types), instructions |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.