Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Carosafe™ Holding Solution
SDPSCI_ED

For use in formalin-preserved specimens. Carolina™ Carosafe™ Holding Solution is formaldehyde-free and cannot be used to fix specimens.

Supplier:  Carolina™ 853343

 View more versions of this product

Catalog No. S37679


Add to Cart

Alert:

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000


Description

Description

  • Carosafe preservative and holding solution for biological specimens contains no formaldehyde and is not a tissue fixative
  • This produces a formalin-preserved specimen that, when dissected, minimizes formaldehyde exposure

Order Info

  • For use in science education laboratories – no certificate of analysis is available
  • Ready-to-use; do not dilute
  • Health Risk: 1
  • Flammability: 1
  • Reactivity: 0

Specifications

Specifications

20 L
Ready to use.
Preservative and holding solution for biological specimens
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit