Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carrageenan, NF, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
500 g
12 kg
Packaging:
Amber Glass Bottle
Poly Bottle
Poly Pail
7/1/9000
7/1/9000

C1107, 9000-07-1

Spectrum™ Carrageenan, NF is used as thickening agent in lotion formulations. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Loss on Drying 12.5%
Quantity 5 g
Packaging Amber Glass Bottle
CAS Max % 100%
Grade NF