Learn More
Alfa Aesar™ CART (62-76), Human, Rat
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J66530MCR
 View more versions of this product
Catalog No. AAJ66530MCR
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
1568.75
Human, Rat
Solid
RUO
CART (62-76), human, rat
HPLC
1mg
Tyr-Gly-Gln-Val-Pro-Met-Cys-Asp-Ala-Gly-Glu-Gln-Cys-Ala-Val (Disulfide bridge: Cys68-Cys74)
-30° to -10°C