Learn More
Cole-Parmer Pentair 155274-03 Mixed-Bed deionization cartridge; 20"L, 0.5 GPM  DFS Item
Manufacturer:  Cole-Parmer 2983064
Deionization Cartridges are designed for deionizing water at 16 MΩ, the mixed-bed cartridges also minimize the TOC (total organic carbon) level. Liquid flows through a bed. Not a double or single open-end cartridge. Compatible with any 9-3/4" housing without a center rod (i.e. 01508-25 or VPN 150068). Not compatible with 01508-40 or 01508-41 (VPN 156017-02 or 156018-02). All materials are FDA-compliant.
Catalog No. 50-632-066