Learn More
Cole-Parmer Pentair Double open end hexametaphosphate crystal cartridge; 10"L  DFS Item
Manufacturer:  Cole-Parmer 2983060
Hexametaphosphate Crystal Cartridge prevents scale and rust buildup — use with water heaters, air conditioning equipment, and many other types of water processing equipment. Food-grade hexametaphosphate dissolves slowly in water to treat scale, corrosion, and iron problems at various flow rates and feed concentrations. All materials are FDA-compliant. Water being treated should have no more than 257 mg/L hardness, with a pH range of 6.5 to 9.0. Maximum temperature is 100°F (38°C). Cartridge is made from polypropylene (PP) shell with a polyester pad prefilter and a PP post filter.
Catalog No. 50-632-062