Learn More
Cole-Parmer Pentair WPX100BB20P Fibrillated polypropylene string-wound cartridge, 100 µm, 20" length  DFS Item
Manufacturer:  Cole-Parmer 2980245
Polypropylene string is resistant to chemical and bacterial attack. Suitable to 165°F (74 °C).
Catalog No. 50-632-264