Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-Carveol, 95%, mixture of isomers, ACROS Organics™

Click to view available options
Quantity:
50g
99-48-9
C10H16O
152.237
MFCD00869995
BAVONGHXFVOKBV-UHFFFAOYSA-N
carveol,p-mentha-6,8-dien-2-ol,p-mentha-1,8-dien-6-ol,1-methyl-4-isopropenyl-6-cyclohexen-2-ol,2-cyclohexen-1-ol, 2-methyl-5-1-methylethenyl,p-mentha-1 6 ,8-dien-2-ol,5-isopropenyl-2-methyl-2-cyclohexen-1-ol,--carveol,l-carveol,2-methyl-5-1-methylethenyl-2-cyclohexen-1-ol
7438
CHEBI:23046
2-methyl-5-prop-1-en-2-ylcyclohex-2-en-1-ol
CC1=CCC(CC1O)C(=C)C
99-48-9
C10H16O
152.237
MFCD00869995
BAVONGHXFVOKBV-UHFFFAOYSA-N
carveol,p-mentha-6,8-dien-2-ol,p-mentha-1,8-dien-6-ol,1-methyl-4-isopropenyl-6-cyclohexen-2-ol,2-cyclohexen-1-ol, 2-methyl-5-1-methylethenyl,p-mentha-1 6 ,8-dien-2-ol,5-isopropenyl-2-methyl-2-cyclohexen-1-ol,--carveol,l-carveol,2-methyl-5-1-methylethenyl-2-cyclohexen-1-ol
7438
CHEBI:23046
2-methyl-5-prop-1-en-2-ylcyclohex-2-en-1-ol
CC1=CCC(CC1O)C(=C)C
Specific Rotation -102° to -119° (20°C, 589nm) (c=1, CHCl3)
Infrared Spectrum Authentic
Quantity 50g
Refractive Index 1.4950 to 1.4970 (20°C, 589nm)
Color Colorless to Yellow
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Beilstein 06, I, 61
Formula Weight 152.24
Physical Form Liquid
Percent Purity ≥94%
Chemical Name or Material (-)-Carveol, 98%
Show More Show Less

EINECSNumber : 202-757-4

RTECSNumber : OS8400000

TSCA : TSCA