Carver™ Lab Presses
Range of five automatic or manual models; clamping force up to 25 tons
Manufacturer: Fred S Carver Inc 38510
 View more versions of this product
Catalog No. 13-872
    N/A
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 
    Specifications
    Product Type Lab Press
    Weight (English) 200 lb.
    Weight (Metric) 90kg
    Normal Force 12 tons
    For Use With (Application) For pressing oils, separating liquids from solids, filtering, plastic molding, compression and crush testing
  • Description & Specifications
  • Accessories
Related Products
Product Content Feedback