Learn More
Carver™ Lab Presses
Range of five automatic or manual models; clamping force up to 25 tons
Manufacturer:  Fred S Carver Inc 4350
 View more versions of this product
Catalog No. 13-872-45
Due to product restrictions, please to purchase or view availability for this product.
Specifications Description & Specifications
Specifications
Standard Benchtop Press
115 lb.
12 tons
For kbr pellet pressing, glove box applications and onsite field testing, palletizing, fluid extraction, pressure forming, destructive testing and other testing procedures
52.1kg
Accessories