Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-Carvone, ≥98.5% (sum of enantiomers, GC), MilliporeSigma™ Supelco™

$34.55 - $225.00

Specifications

Percent Purity ≥98.5% (sum of enantiomers, GC)
Density 0.960 g/mL (at 20°C)
Grade Analytical Standard
Linear Formula C10H14 O
Physical Form Neat
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-101-3586
View Documents
MilliporeSigma Supelco
220701ML
1 mL
Each for $34.55
Add to cart
 
11-101-3584
View Documents
MilliporeSigma Supelco
2207025ML
25 mL
Each for $63.10
Add to cart
 
11-101-3585
View Documents
MilliporeSigma Supelco
22070100ML
100 mL
Each for $225.00
Add to cart
 
Description

Description

Featured Industries: Agriculture and Food and Beverages
Specifications

Specifications

≥98.5% (sum of enantiomers, GC)
Analytical Standard
Neat
0.960 g/mL (at 20°C)
C10H14 O
Safety and Handling

Safety and Handling

ShelfLife : Limited shelf life, expiry date on the label

SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit