Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cas9 Monoclonal Antibody (6F7), Invitrogen™

Mouse Monoclonal Antibody

Supplier:  Thermo Scientific MA547461

Encompass

Catalog No. PIMA547461


Add to Cart

Description

Description

Cas9 Monoclonal antibody specifically detects Cas9 in Bacteria samples. It is validated for Immunocytochemistry, Immunohistochemistry, Western Blot.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cas9
Monoclonal
1 mg/mL
PBS with 50% glycerol and 5mM sodium azide
Mouse
Protein A/G
RUO
Bacteria
Antibody
IgG1
Immunocytochemistry, Immunohistochemistry, Western Blot
6F7
Unconjugated
cas9; CASP 9; Caspase9; Caspase-9 precursor; CASPASE-9c; CRISPR9; CRISPR-Cas9; CTD; OTTHUMP00000002324; RNCASP9; RP11-265F14.3; s aureus
C-terminal region of S. aureus, amino acids 803-1053 of sequence expressed in and purified from E. coli.
100 μL
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit