Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Casanthranol, USP, 20%, Spectrum™

Click to view available options
Quantity:
100 g
500 g
25 kg
Packaging:
Fiber Drum
Poly Bottle
8024-48-4
8024-48-4

C1101, 8024-48-4

Spectrum™ Casanthranol, USP is used as a laxative. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Quantity 500 g
Ignition Residue 4%
Loss on Drying 10%
Packaging Poly Bottle
CAS Max % 100%
Percent Purity 20%