Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CASC3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555116
Description
Immunogen sequence: DAPVLGSPEK EEAASEPPAA APDAAPPPPD RPIEKKSYSR ARRTRTKVGD AVKLAEEVPP PPEGLIPAPP VPETTPTPPT KTGTWEAPVD SSTSGLEQDV AQLN Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 81%.
The 26S proteasome is a multicatalytic proteinase complex with a highly ordered structure composed of 2 complexes, a 20S core and a 19S regulator. The 20S core is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. The 19S regulator is composed of a base, which contains 6 ATPase subunits and 2 non-ATPase subunits, and a lid, which contains up to 10 non-ATPase subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. The immunoproteasome contains an alternate regulator, referred to as the 11S regulator or PA28, that replaces the 19S regulator. Three subunits of the 11S regulator have been identified. This gene encodes the gamma subunit of the 11S regulator. Six gamma subunits combine to form a homohexameric ring. Two transcript variants encoding different isoforms have been identified.
Specifications
CASC3 | |
Polyclonal | |
Unconjugated | |
CASC3 | |
Barentsz; barentsz protein; BTZ; cancer susceptibility candidate 3; cancer susceptibility candidate gene 3 protein; Cancer susceptibility candidate gene 3 protein homolog; Casc3; mBtz; metastatic lymph node 51; metastatic lymph node gene 51 protein; Metastatic lymph node gene 51 protein homolog; MLN 51; MLN 51 homolog; Mln51; Protein barentsz; protein CASC3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22794 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O15234 | |
CASC3 | |
Recombinant protein corresponding to Human CASC3. Recombinant protein control fragment (Product #RP-91838). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction