Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cascara Sagrada Fluidextract, USP, Spectrum™

Click to view available options
Quantity:
500 mL
4 L
Packaging:
Amber Glass Bottle

CA202

Spectrum™ Cascara Sagrada Fluidextract, USP - All Spectrum Chemical manufactured USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Quantity 4 L
Packaging Amber Glass Bottle
CAS Max % 100%
UN Number UN1170