Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Casein Blocking Buffer, 3% in PBS, with 0.02% sodium azide, Thermo Scientific™

For Western blots and ELISA

Manufacturer:  Thermo Scientific Chemicals J63453AP

 View more versions of this product

Catalog No. AAJ63453AP

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

500 mL
Liquid
-30°C to -10°C
Western blots and ELISA
Blocking Buffer
Liquid
Safety and Handling

Safety and Handling

Recommended Storage : Store at -20°C

SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit