Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Affymetrix/USB™ Casein, High Nitrogen

Prepared from New Zealand casein exclusively.

$96.40 - $381.00

Specifications

Format Powder
Name Casein
Packaging Plastic bottle
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ12845P5 View Documents Affymetrix/USB™
J12845P5
2.2kg (5 lb.) Each for $96.40
Order in Bulk

Request bulk or custom formats

Add to cart
 
AAJ12845Q1 View Documents Affymetrix/USB™
J12845Q1
11.3kg (25 lb.) Each for $381.00
Order in Bulk

Request bulk or custom formats

Add to cart
 
Description

Description

  • Format: White to off-white powder
  • Protein (Dry basis): ≥95.0%
  • Melting point: ~280°C (dec.)
  • MDLNumber: MFCD00081481
  • CAS: 9000-71-9
Specifications

Specifications

Powder
Casein
Plastic bottle
Safety and Handling

Safety and Handling

EINECSNumber : 232-555-1

Recommended Storage : Ambient temperatures; Store under Nitrogen

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit