Learn More
Casein hydrolysate enzymatic, Alfa Aesar™
Used as nutritional source in microbial media. It is derived from the enzymatic digest of casein, often used in culture media. NZ Amine EKC contains a mixture of peptides and amino acids.
Manufacturer:  Alfa Aesar J6456522
 View more versions of this product
Catalog No. AAJ6456522
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
Powder
100g