Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Casein Kinase 1 epsilon Antibody (OTI5D4), DyLight 488, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP270464G

Catalog No. NBP270464G


Add to Cart

Description

Description

Casein Kinase 1 epsilon Monoclonal specifically detects Casein Kinase 1 epsilon in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

Casein Kinase 1 epsilon
Monoclonal
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
CSNK1E
Recombinant protein fragment corresponding to amino acids 153-416 of human Casein Kinase 1 epsilon (NP_689407) produced in E.coli.
0.1 mL
Primary
Human, Mouse, Rat
IgG1
Western Blot, Immunohistochemistry
OTI5D4
Western Blot, Immunohistochemistry
casein kinase 1, epsilon, casein kinase I isoform epsilon, CKIe, CKI-epsilon, EC 2.7.11, EC 2.7.11.1, HCKIE, MGC10398
Mouse
Affinity Purified
Protein Kinase, Wnt Signaling Pathway
1454.0
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit