Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Casein Kinase 1 epsilon Mouse anti-Human, Janelia Fluor™ 646, Clone: OTI5D4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270464JF646

Catalog No. N270464J646


Add to cart

Description

Description

Casein Kinase 1 epsilon Monoclonal antibody specifically detects Casein Kinase 1 epsilon in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

Casein Kinase 1 epsilon
OTI5D4
Immunohistochemistry
Affinity Purified, Conjugated
CSNK1E
Recombinant protein fragment corresponding to amino acids 153-416 of human Casein Kinase 1 epsilon (NP_689407) produced in E.coli.
Immunogen affinity purified
Protein Kinase, Wnt Signaling Pathway
Primary
1454.0
Immunohistochemistry
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
casein kinase 1, epsilon, casein kinase I isoform epsilon, CKIe, CKI-epsilon, EC 2.7.11, EC 2.7.11.1, HCKIE, MGC10398
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit