Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Casein Kinase 1 epsilon Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen Casein Kinase 1 epsilon
Applications Immunofluorescence, Immunohistochemistry, Immunoprecipitation, KnockDown, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB092435
View Documents
Novus Biologicals
NBP303608100UL
100 μg Each for $320.71
Add to cart
 
NB092436
View Documents
Novus Biologicals
NBP30360820UL
20 μg Each for $138.37
Add to cart
 
Description

Description

Casein Kinase 1 epsilon Polyclonal antibody specifically detects Casein Kinase 1 epsilon in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, KnockDown
Specifications

Specifications

Casein Kinase 1 epsilon
Unconjugated
Rabbit
Protein Kinase, Wnt Signaling Pathway
PBS with 50% glycerol, pH7.3.
1454
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, KnockDown, Western Blot
Purified
RUO
Human, Mouse, Rat
casein kinase 1, epsilon, casein kinase I isoform epsilon, CKIe, CKI-epsilon, EC 2.7.11, EC 2.7.11.1, HCKIE, MGC10398
Recombinant fusion protein containing a sequence corresponding to amino acids 247-416 of human Casein Kinase 1 epsilon (NP_689407.1).
Polyclonal
Affinity purified
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit