Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Casein Kinase 1 gamma 2 Antibody (OTI4H3), Alexa Fluor™ 532, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP270465AF532

Catalog No. NP27046A32


Only null left
Add to Cart

Description

Description

Casein Kinase 1 gamma 2 Monoclonal specifically detects Casein Kinase 1 gamma 2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

Casein Kinase 1 gamma 2
Monoclonal
Alexa Fluor 532
50 mM sodium borate with 0.05% sodium azide
CSNK1G2
Recombinant protein fragment corresponding to amino acids 163-415 of human Casein Kinase 1 gamma 2 (NP_001310) produced in E.coli.
0.1 mL
Primary
Human, Mouse, Rat
IgG2a
Western Blot, Immunohistochemistry
OTI4H3
Western Blot, Immunohistochemistry
casein kinase 1 isoform gamma-2, casein kinase 1, gamma 2, casein kinase I isoform gamma-2, CK1g2, CKI-gamma 2, EC 2.7.11, EC 2.7.11.1
Mouse
Affinity Purified
Protein Kinase
1455.0
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.