Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Casein Kinase 1 gamma 2 Mouse anti-Human, Alexa Fluor™ 700, Clone: OTI4H3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270465AF700

Catalog No. N270465A700


Add to cart

Description

Description

Casein Kinase 1 gamma 2 Monoclonal antibody specifically detects Casein Kinase 1 gamma 2 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

Casein Kinase 1 gamma 2
OTI4H3
Western Blot, Immunohistochemistry
Affinity Purified, Conjugated
CSNK1G2
Recombinant protein fragment corresponding to amino acids 163-415 of human Casein Kinase 1 gamma 2 (NP_001310) produced in E.coli.
Immunogen affinity purified
Protein Kinase
Primary
1455.0
Immunohistochemistry, Western Blot
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
casein kinase 1 isoform gamma-2, casein kinase 1, gamma 2, casein kinase I isoform gamma-2, CK1g2, CKI-gamma 2, EC 2.7.11, EC 2.7.11.1
Mouse
IgG2a
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit