Learn More
Casein Kinase 2 beta Sheep anti-Human, Mouse, Rat, Polyclonal, R&D Systems™
Sheep Polyclonal Antibody
Manufacturer:  R&D Systems AF4380SP
Catalog No. AF4380SP
Add to cart
This item is not returnable. View return policy
Description
Description
Casein Kinase 2 beta Polyclonal antibody AF4380SP specifically detects Casein Kinase 2 beta in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western Blotting
Specifications
Specifications
Casein Kinase 2 beta
LYOPH
Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL
P67870
casein kinase 2, beta polypeptide, Casein kinase II beta subunit, casein kinase II subunit beta, CK II beta, CK2B, CK2N, CSK2B, CSNK2B, G5A, MGC138222, MGC138224, phosvitin, Protein G5a
Sheep
IgG
25 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human, mouse and rat Casein Kinase 2 beta in Western blots.
Human, Mouse, Rat
Western Blot, Immunocytochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
CSNK2B
E. coli-derived recombinant human Casein Kinase 2 beta Met1-His153 Accession # P67870
Antigen Affinity-purified
RUO
Primary
1460
Reconstitute at 0.2 mg/mL in sterile PBS.