Learn More
Alfa Aesar™ Casein Sodium Salt
For research use
Manufacturer:  Alfa Aesar™ J65590A1
 View more versions of this product
Catalog No. AAJ65590A1
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
Casein is frequently used as a blocking agent in immunochemistry experiments. It is also used in the recovery of enzyme activity from SDS extracted samples, and as a substrate for protease and kinase assays.
Lyophilized powder
Bovine milk
Casein sodium salt
Lyophilized Powder
1kg
RUO
No