Learn More
Casella™ CEL-24X Digital Sound Level Meters
Choice of four meters to meet your workplace noise monitoring needs
Manufacturer:  CEL Instruments LtdSupplier Diversity Partner CEL244/K2
 View more versions of this product
Catalog No. 19-979-639
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
1KHZ
1.2 in.
LCD
8.3 in.
30 to 90%
30 to 130dB
CEL-244 PC kit
0° to 40°C
8.8 oz.
2.8 in.
ANSI S1.4, IEC 616672 Type 2, ANSI S1.43
3.04cm
70dB
21.08cm
Foam windscreen, three AA batteries, wrist strap, meter, single level acoustic calibrator, small kit case, USB cable to PC, dB24 software on a CD
<33dB
32° to 104°F
2 Years
249.47g
7.11cm