Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CASK Interacting Protein 1 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP281707MFV610
Description
CASK Interacting Protein 1 Polyclonal antibody specifically detects CASK Interacting Protein 1 in Human samples. It is validated for Western Blot, ELISASpecifications
CASK Interacting Protein 1 | |
Polyclonal | |
Western Blot, ELISA | |
ANKS5A, caskin-1, CASKIN1, CASK-Interacting Protein 1, KIAA1306 | |
CASK Interacting Protein 1 antibody was raised against a 17 amino acid synthetic peptide near the center of human CASK Interacting Protein 1. The immunogen is located within amino acids 720 - 770 of CASK Interacting Protein 1. Amino Acid Squence: TPPREARPGRHGHSIKR | |
0.1 mL | |
Neuroscience | |
57524 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction