Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CASKIN2 Monoclonal Antibody (OTI4A3), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene CF812217


Description
CASKIN2 Monoclonal antibody specifically detects CASKIN2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
CASKIN2 | |
Monoclonal | |
See Label | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P20722, P22004, Q04906 | |
Mouse | |
Protein A/G | |
RUO | |
12161, 25644, 654 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2b |
Western Blot | |
OTI4A3 | |
Unconjugated | |
BMP6 | |
1600028L06Rik; ANKS5B; CASK interacting protein 2; Caskin2; Caskin-2; cask-interacting protein 2; KIAA1139; mKIAA1139 | |
Human recombinant protein fragment corresponding to amino acids 21-374 of human BMP6 (NP_001709) produced in E.coli. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction