Learn More
CASP4 Mouse anti-Human, Clone: OTI9F6, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF805591
Catalog No. 50-166-7495
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Caspases are cysteine proteases, expressed as inactive precursors, that mediate apoptosis by proteolysis of specific substrates. Caspases have the ability to cleave after aspartic acid residues. There are two classes of caspases involved in apoptosis; initiators (activation by receptor cluster) and effectors (activation by mitochondrial permeability transition). Proapoptotic signals autocatalytically activate initiator caspases, such as Caspase-8 and Caspase 9. Activated initiator caspases then process effector caspases, such as Caspase-3 and Caspase-7, which in turn cause cell collapse.
Specifications
Specifications
CASP4
Monoclonal
Unconjugated
CASP4
Lyophilized
CASP4
Human recombit protein fragment corresponding to amino acids 81-270 of human CASP4 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI9F6
PBS with 8% trehalose and no preservative; pH 7.3
P49662
ICE(rel)II, ICEREL-II, ICH-2, Mih1, Mih1/TX, TX
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
837