Learn More
Abnova™ CASP6 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00000839T02
Catalog No. 89-938-095
Add to cart
Description
Description
Specifications
Specifications
Western Blot
CASP6 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 33.3kDa
Transient overexpression cell lysate was tested with anti-CASP6 antibody (Mfr. No. H00000839-D01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
293T
NM_001226
pCMV-CASP6 full-length
100 μL
Human