Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CASP7 Mouse anti-Human, Clone: 7-1-11, Abnova™

Mouse monoclonal antibody raised against human CASP7.

Manufacturer:  Abnova Corporation MAB12590

Catalog No. 89-933-726


Add to cart

Description

Description

This gene encodes a protein which is a member of the cysteine-aspartic acid protease (caspase) family. Sequential activation of caspases plays a central role in the execution-phase of cell apoptosis. Caspases exist as inactive proenzymes which undergo proteolytic processing at conserved aspartic residues to produce two subunits, large and small, that dimerize to form the active enzyme. The precursor of this caspase is cleaved by caspase 3 and 10. It is activated upon cell death stimuli and induces apoptosis. Alternative splicing results in four transcript variants, encoding three distinct isoforms. [provided by RefSeq]

Specifications

Specifications

CASP7
7-1-11
Mouse monoclonal antibody raised against human CASP7.
In PBS, 10mM Na3PO4, 50mM NaCl, pH 7.2 (0.1mM PMSF)
CMH-1/ICE-LAP3/MCH3
Mouse
IgG
50 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot The optimal working dilution should be determined by the end user.
Liquid
CASP7
Recombinant protein corresponding to full length human CASP7.
Protein G purification
RUO
Primary
840
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit