Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caspase 1 Monoclonal Antibody (B0-6), Invitrogen™

Catalog No. PIMA532909
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIMA532909 100 μL
1 options

Catalog No. PIMA532909

Supplier: Thermo Scientific MA532909

Mouse Monoclonal Antibody

Caspase 1 Monoclonal antibody specifically detects Caspase 1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Caspase 1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone B0-6
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Gene CASP1
Gene Accession No. P29452, P29466, P43527
Gene Alias Casp1; CASP-1; Caspase-1; ICE; IL1BC; IL-1BC; IL1BCE; Interleukin-1 beta convertase; Interleukin-1 beta-converting enzyme; P45
Gene Symbols CASP1
Host Species Mouse
Immunogen Recombinant protein with Human Caspase-1 aa 126-333
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12362, 25166, 834
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.