Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caspase-1 (p20) Rabbit anti-Human,Mouse,Rat, KHL, Clone: 2G4, MilliporeSigma™

Rabbit Monoclonal Antibody

Specifications

Antigen Caspase-1 (p20)
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Conjugate KHL
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ZRB123325MI
View Documents
MilliporeSigma
ZRB123325UL
25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
ZRB12334XMI
View Documents
MilliporeSigma
ZRB12334X25UL
4 x 25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Caspase-1 (p20) Monoclonal antibody specifically detects Caspase-1 (p20) in Human,Mouse,Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blotting.
Specifications

Specifications

Caspase-1 (p20)
KHL
Rabbit
EC:3.4.22.36;CASP-1;Interleukin-1 beta convertase;IL-1BC;Interleukin-1 beta-converting enzyme;ICE;IL-1 beta-converting enzyme;p45
KLH-conjugated linear peptide corresponding to 21 amino acids from the p20 subunit of human Caspase-1.
Primary
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Purified
Human, Mouse, Rat
CASP1, IL1BC, IL1BCE
Monoclonal
Protein A Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit