Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Caspase-11, Rat anti-Mouse,Rat, Clone: 400000000000, Novus Biologicals
Rat Monoclonal Antibody
Manufacturer: Novus Biologicals NBP280100
Description
Caspase-11 Monoclonal antibody specifically detects Caspase-11 in Mouse, Rat Samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunoprecipitation.Specifications
Caspase-11 | |
4e+011 | |
Unconjugated | |
PBS | |
ich-3, CASP11, CASP-11, CASP4, CASP-4, caspase 4, apoptosis-related cysteine peptidase, Caspl | |
Rat | |
IgG1 Kappa | |
100 ug | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
Monoclonal | |
Recognizes mouse and rat full-length Caspase-11 protein. Detects two bands of ∽38kDa and ∽43kDa by Western blot. |
ELISA, Flow Cytometry, Immunoprecipitation, Western Blot | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunoprecipitation | |
Purified | |
Casp4 | |
Recombinant mouse Caspase-11 (p20 fragment). | |
Protein G purified | |
RUO | |
Primary | |
12363 | |
Mouse, Rat |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title