Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Caspase-3 Colorimetric Assay Kit

Caspase-3/CPP32 Colorimetric Assay Kit provides a simple and convenient means for assaying the activity of caspases that recognize the sequence DEVD.

Manufacturer:  R&D Systems™ K106100

Catalog No. K106100


Add to cart
This item is not returnable. View return policy

Description

Description

Kit Contents:

  • Cell Lysis Buffer (100 mL)
  • 2X Reaction Buffer (4 x 2 mL)
  • DEVD-pNA (4 mM) (0.5 mL)
  • DTT (1 M) (0.4 mL)
  • Dilution Buffer (100 mL)
Specifications

Specifications

100 Tests
Store the unopened product at -20 to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit