Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Caspase 3 (CPP32) Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation procedures.

Specifications

Antigen Caspase 3 (CPP32) Ab-1
Clone 3CSP01; same as 7.1.44
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 1
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS1121P1
View Documents
Thermo Scientific™
MS1121P1
Purified Ab with BSA and Azide 500μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation procedures with Thermo Scientific™ Caspase 3 (CPP32) Ab-1, Mouse Monoclonal Antibody.

Caspase 3 is ubiquitously expressed and like other caspases is synthesized as an inactive (32 kDa) proenzyme. Upon activation, Caspase 3 is cleaved at Asp28-Ser29 and Asp175-Ser176 thereby generating two subunits of 17kDa and 12kDa, respectively. Recent studies have implicated that Caspase 3 is associated with the induction of apoptosis. Activation of Caspase 3 occurs in response to a variety of apoptotic inducers including Fas-mediated apoptosis.

Host Species: Mouse

Clone: 3CSP01; same as 7.1.44

Isotype: IgG2a

Species Reactivity: Human. Others not tested.

Epitope: Not determined

Immunogen: Recombinant full-length human Caspase 3 protein

Molecular Weight: 32 kDa

Positive Control: Jurkat cells, Tonsil and appendix

Cellular Localization: Predominantly Cytoplasmic with some nuclear staining

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Caspase 3 (CPP32) Ab-1
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
Murine
Apoptosis
Recombinant full length human Caspase 3 protein
3CSP01; same as 7.1.44
Monoclonal
Concentrated
RUO
Primary
IgG2a
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit