Learn More
Alfa Aesar™ Caspase-3 Inhibitor I, cell-permeable
Manufacturer:  Alfa Aesar™ J66300EXD
 View more versions of this product
Catalog No. AAJ66300EXD
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
0.5mg
−30° to −10°C
C94H158N20O27
N-Acetyl-Ala-Ala-Val-Ala-Leu-Leu-Pro-Ala-Val-Leu-Leu-Ala-Leu-Leu-Ala-Pro-Asp-Glu-Val-Asp-al; Ac-A-A-V-A-L-L-P-A-V-L-L-A-L-L-A-P-D-E-V-D-CHO
White to Off-white
Lyophilized Powder
2000.37g/mol
Signal Transduction Reagent